3-Phenylpropylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
4,4,5,5-Tetramethyl-2-(3-phenylpropyl)-1,3,2-dioxaborolane, 3-Phenyl-1-propylboronic acid pinacol ester
H28985-03
H28985-06
AAH28985-03EA
349
USD
AAH28985-03
AAH28985-06
AAH28985-MD
3-Phenylpropylboronic acid pinacol ester 97%
3-Phenylpropylboronic acid pinacol ester
Purity: 97%
CAS No.: 329685-40-7
Molecular Formula: C15H23BO2
Formula Weight: 246.16
MDL No.: MFCD09953505
Formula:
C₁₅H₂₃BO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09953505 CAS Number: 329685-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...