Potassium (4-iodophenyl)trifluoroborate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Iodophenyl)trifluoroboric acid potassium salt, Potassium 4-iodophenyltrifluoroborate
H28758-03
H28758-06
AAH28758-03EA
82
USD
AAH28758-03
AAH28758-06
Potassium (4-iodophenyl)trifluoroborate 96%
Potassium (4-iodophenyl)trifluoroborate
Purity: 96%
CAS No.: 912350-00-6
Molecular Formula: C6H4BF3IK
Formula Weight: 309.90
Melting Point: 297-299°
MDL No.: MFCD09800738
Formula:
C₆H₄BF₃IK Melting Pt: 297…299 °C Storage Temperature: Ambient |
MDL Number:
MFCD09800738 CAS Number: 912350-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...