3-(4-Methoxyphenyl)propylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester

H28557-03 H28557-06 H28557-MD
AAH28557-03EA 137 USD
AAH28557-03 AAH28557-06 AAH28557-MD
3-(4-Methoxyphenyl)propylboronic acid pinacol ester 97%
3-(4-Methoxyphenyl)propylboronic acid pinacol ester

Purity: 97% Molecular Formula: C16H25BO3 Formula Weight: 276.19 MDL No.: MFCD09953503

Formula: C₁₆H₂₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD09953503
CAS Number: 1073371-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR