3-Aminophenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Aminobenzeneboronic acid pinacol ester
H28448-03
H28448-06
AAH28448-03EA
83.4
USD
AAH28448-03
AAH28448-06
3-Aminophenylboronic acid pinacol ester 97%
3-Aminophenylboronic acid pinacol ester
Purity: 97%
CAS No.: 210907-84-9
Molecular Formula: C12H18BNO2
Formula Weight: 219.09
Melting Point: 87-93°
MDL No.: MFCD03453668
Formula:
C₁₂H₁₈BNO₂ MW: 219.09 Melting Pt: 87…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD03453668 CAS Number: 210907-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...