2,6-Dimethoxynaphthalene 99%
Supplier: Thermo Scientific Chemicals
H28366-03
H28366-06
AAH28366-03EA
25.7
USD
AAH28366-03
AAH28366-06
2,6-Dimethoxynaphthalene 99%
2,6-Dimethoxynaphthalene
Purity: 99%
CAS No.: 5486-55-5
Molecular Formula: C12H12O2
Formula Weight: 188.23
Melting Point: 152-154°
MDL No.: MFCD00004063
Formula:
C₁₂H₁₂O₂ MW: 188.23 g/mol Melting Pt: 152…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004063 CAS Number: 5486-55-5 EINECS: 226-816-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...