4-Chloro-2-(trifluoromethyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-2-(trifluoromethyl)benzeneboronic acid, (4-Chloro-2-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-chloro-2-(trifluoromethyl)phenyl)borane

H28351-03 H28351-09
AAH28351-03EA 84.9 USD
AAH28351-03 AAH28351-09
4-Chloro-2-(trifluoromethyl)phenylboronic acid 97%
4-Chloro-2-(trifluoromethyl)phenylboronic acid

Purity: 97% CAS No.: 313545-41-4 Molecular Formula: C7H5BClF3O2 Formula Weight: 224.38 Melting Point: 180-182° MDL No.: MFCD04973086

Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD04973086
CAS Number: 313545-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR