1,7-Dihydroxynaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Naphthalene-1,7-diol

H28261-14 H28261-06
AAH28261-14EA 170 USD
AAH28261-14 AAH28261-06
1,7-Dihydroxynaphthalene 97%
1,7-Dihydroxynaphthalene

Purity: 97% CAS No.: 575-38-2 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 180-184° MDL No.: MFCD00035720

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00035720
CAS Number: 575-38-2
EINECS: 209-383-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR