3-Iodophenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Iodobenzeneboronic acid pinacol ester, 2-(3-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H28115-03
H28115-06
AAH28115-03EA
47.8
USD
AAH28115-03
AAH28115-06
3-Iodophenylboronic acid pinacol ester 97%
3-Iodophenylboronic acid pinacol ester
Purity: 97%
CAS No.: 408492-28-4
Molecular Formula: C12H16BIO2
Formula Weight: 329.97
Melting Point: 70-72°
MDL No.: MFCD09953498
Formula:
C₁₂H₁₆BIO₂ MW: 329.97 g/mol Melting Pt: 70…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD09953498 CAS Number: 408492-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...