4-Amino-m-anisic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-3-methoxybenzenecarboxylic acid

H28098-03 H28098-06
AAH28098-03EA 74 USD
AAH28098-03 AAH28098-06
4-Amino-m-anisic acid 95%
4-Amino-m-anisic acid

Purity: 95% CAS No.: 2486-69-3 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: 186-188° MDL No.: MFCD00016539

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 186…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00016539
CAS Number: 2486-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR