4-Bromo-2-methylpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27884-03
H27884-MD
AAH27884-03EA
92.5
USD
AAH27884-03
AAH27884-MD
4-Bromo-2-methylpyridine 97%
4-Bromo-2-methylpyridine
Purity: 97%
CAS No.: 22282-99-1
Molecular Formula: C6H6BrN
Formula Weight: 172.03
Flash Point: 79°(174°F)
Refractive Index: 1.556
Odor: Characteristic
Boiling Point: 76°/14mm
MDL No.: MFCD03788196
Formula:
C₆H₆BrN Boiling Pt: 76 °C/14 mm Density: 1.45 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03788196 CAS Number: 22282-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...