(2-Bromo-4-methoxyphenyl)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H27859-14 H27859-03 H27859-06
AAH27859-14EA 303 USD
AAH27859-14 AAH27859-03 AAH27859-06
(2-Bromo-4-methoxyphenyl)acetic acid 97%
(2-Bromo-4-methoxyphenyl)acetic acid

Purity: 97% CAS No.: 66916-99-2 Molecular Formula: C9H9BrO3 Formula Weight: 245.07 Melting Point: 127-131° MDL No.: MFCD02664700

Formula: C₉H₉BrO₃
Melting Pt: 127…131 °C
Storage Temperature: Ambient
MDL Number: MFCD02664700
CAS Number: 66916-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR