2,4,5-Trifluorophenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27858-06
H27858-14
AAH27858-06EA
151
USD
AAH27858-06
AAH27858-14
2,4,5-Trifluorophenylboronic acid 97%
2,4,5-Trifluorophenylboronic acid
Purity: 97%
CAS No.: 247564-72-3
Molecular Formula: C6H4BF3O2
Formula Weight: 175.90
Melting Point: 99-101°
MDL No.: MFCD01863165
Formula:
C₆H₄BF₃O₂ MW: 175.9 g/mol Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863165 CAS Number: 247564-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...