4-Hydroxyphenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 4-Hydroxybenzeneboronic acid pinacol ester
H27836-06
H27836-03
AAH27836-06EA
243
USD
AAH27836-06
AAH27836-03
4-Hydroxyphenylboronic acid pinacol ester 97%
4-Hydroxyphenylboronic acid pinacol ester
Purity: 97%
CAS No.: 269409-70-3
Molecular Formula: C12H17BO3
Formula Weight: 220.07
Melting Point: 114-117°
MDL No.: MFCD02093756
Formula:
C₁₂H₁₇BO₃ MW: 220.08 g/mol Melting Pt: 114…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093756 CAS Number: 269409-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...