1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Butyl-2,3-dimethyl-1H-imidazol-3-ium hexafluorophosphate(V)
H27827-18
H27827-06
AAH27827-18EA
290
USD
AAH27827-18
AAH27827-06
1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate 99%
1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate
Purity: 99%
CAS No.: 227617-70-1
Molecular Formula: C9H17F6N2P
Formula Weight: 298.21
Refractive Index: 1.423
Melting Point: 48-50°
MDL No.: MFCD03790877
Formula:
C₉H₁₇F₆N₂P MW: 298.21 g/mol Melting Pt: 48…50 °C Density: 1.354 Storage Temperature: Ambient |
MDL Number:
MFCD03790877 CAS Number: 227617-70-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...