3-Cyano-4-pyridineboronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Cyanopyridine-4-boronic acid, pinacol ester
H27814-03
H27814-MD
AAH27814-03EA
361
USD
AAH27814-03
AAH27814-MD
3-Cyano-4-pyridineboronic acid pinacol ester 95%
3-Cyano-4-pyridineboronic acid pinacol ester
Purity: 95%
CAS No.: 878194-92-4
Molecular Formula: C12H15BN2O2
Formula Weight: 230.07
Melting Point: 97-99°
MDL No.: MFCD09037481
Formula:
C₁₂H₁₅BN₂O₂ MW: 230.07 g/mol Melting Pt: 97…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037481 CAS Number: 878194-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...