4-Nitrophenylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Nitrobenzeneboronic acid, (4-Nitrophenyl)boronic acid, Dihydroxy(4-nitrophenyl)borane, (4-Nitrophenyl)metaboric acid

H27767-03 H27767-06
AAH27767-03EA 69.5 USD
AAH27767-03 AAH27767-06
4-Nitrophenylboronic acid 95%
4-Nitrophenylboronic acid

Purity: 95% CAS No.: 24067-17-2 Molecular Formula: C6H6BNO4 Formula Weight: 166.93 Melting Point: ca 285° dec. MDL No.: MFCD00161360

Formula: C₆H₆BNO₄
MW: 166.93 g/mol
Melting Pt: ∼ 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00161360
CAS Number: 24067-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR