1-Butyl-3-methylimidazolium methyl sulphate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
[BMIM] [MeSO4], 1-n-Butyl-3-methylimidazolium methyl sulphate, ECOENG 411, BMIMMeSO4
H27754-18
H27754-06
AAH27754-18EA
220
USD
AAH27754-18
AAH27754-06
1-Butyl-3-methylimidazolium methyl sulphate 99%
1-Butyl-3-methylimidazolium methyl sulphate
Purity: 99%
CAS No.: 401788-98-5
Molecular Formula: C9H18N2O4S
Formula Weight: 250.32
Refractive Index: 1.4780
Formula:
C₉H₁₈N₂O₄S Density: 1.21 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03095437 CAS Number: 401788-98-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...