1-Butyl-3-methylimidazolium octylsulfate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27720-18
H27720-06
AAH27720-18EA
400
USD
AAH27720-18
AAH27720-06
1-Butyl-3-methylimidazolium octylsulfate 99%
1-Butyl-3-methylimidazolium octylsulphate
Purity: 99%
CAS No.: 445473-58-5
Molecular Formula: C16H32N2O4S
Formula Weight: 348.50
MDL No.: MFCD03788914
Formula:
C₁₆H₃₂N₂O₄S Storage Temperature: Ambient |
MDL Number:
MFCD03788914 CAS Number: 445473-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...