1-Methyl-3-propylimidazolium iodide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Methyl-3-n-propylimidazolium iodide, 1-Methyl-3-propyl-1H-imidazol-3-ium iodide
H27682-18
H27682-06
AAH27682-18EA
233
USD
AAH27682-18
AAAH27682-06
1-Methyl-3-propylimidazolium iodide 98%
1-Methyl-3-propylimidazolium iodide
Purity: 98%
CAS No.: 119171-18-5
Molecular Formula: C7H13IN2
Formula Weight: 252.09
Flash Point:
Formula:
C₇H₁₃IN₂ MW: 252.1 g/mol Boiling Pt: 301…304 °C Density: 1.54 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD08276385 CAS Number: 119171-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...