8-Bromo-1-octanol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 8-Bromooctan-1-ol, 1-Bromo-8-hydroxyoctane

H27628-03 H27628-06 H27628-14
AAH27628-03EA 52 USD
AAH27628-03 AAH27628-06 AAH27628-14
8-Bromo-1-octanol 95%
8-Bromo-1-octanol

Purity: 95% CAS No.: 50816-19-8 Molecular Formula: C8H17BrO Formula Weight: 209.13 Flash Point: 110°(230°F) Refractive Index: 1.4810 Boiling Point: 79-80°/0.07mm Beilstein Reference: 1900837 MDL No.: MFCD00010388

Formula: BrH₂C(CH₂)₆CH₂OH
MW: 209.13 g/mol
Boiling Pt: 79…80 °C/0.07 mm
Density: 1.220
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010388
CAS Number: 50816-19-8
EINECS: 256-785-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR