6-Fluoropyridine-2-carbonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Fluoropicolinonitrile, 2-Cyano-6-fluoropyridine

H27600-06 H27600-MD H27600-03
AAH27600-06EA 778 USD
AAH27600-06 AAH27600-MD AAH27600-03
6-Fluoropyridine-2-carbonitrile 95%
6-Fluoropyridine-2-carbonitrile

Purity: 95% CAS No.: 3939-15-9 Molecular Formula: C6H3FN2 Formula Weight: 122.10 Melting Point: 32-35° MDL No.: MFCD09263323

Formula: C₆H₃FN₂
MW: 122.1 g/mol
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD09263323
CAS Number: 3939-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR