2-Chloro-4-fluorophenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H27591-03 H27591-06 H27591-14
AAH27591-03EA 94 USD
AAH27591-03 AAH27591-06 AAH27591-14
2-Chloro-4-fluorophenylboronic acid 98%
2-Chloro-4-fluorophenylboronic acid

Purity: 98% CAS No.: 313545-72-1 Molecular Formula: C6H5BClFO2 Formula Weight: 174.37 MDL No.: MFCD02684295

Formula: C₆H₅BClFO₂
Storage Temperature: Ambient
MDL Number: MFCD02684295
CAS Number: 313545-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR