1-Ethyl-3-methylimidazolium ethylsulphate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: EMIM ESU

H27571-18 H27571-30
AAH27571-18EA 102 USD
AAH27571-18 AAH27571-30
1-Ethyl-3-methylimidazolium ethylsulphate 99%
1-Ethyl-3-methylimidazolium ethylsulphate

Purity: 99% CAS No.: 342573-75-5 Molecular Formula: C8H16N2O4S Formula Weight: 236.29 Flash Point: 162°(324°F) Melting Point: -30° MDL No.: MFCD06798189

Formula: C₈H₁₆N₂O₄S
Melting Pt: <–30 °C
Density: 1.24
Flash Pt: 162 °C(324 °F)
Storage Temperature: Ambient
MDL Number: MFCD06798189
CAS Number: 342573-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR