5-Amino-2-methoxypyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H27528-14 H27528-06
AAH27528-14EA 173 USD
AAH27528-14 AAH27528-06
5-Amino-2-methoxypyridine 98%
5-Amino-2-methoxypyridine

Purity: 98% CAS No.: 6628-77-9 Molecular Formula: C6H8N2O Formula Weight: 124.14 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5735 Melting Point: 29-31° Boiling Point: 85-90°/1mm MDL No.: MFCD00006264

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 28 - 30 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006264
CAS Number: 6628-77-9
EINECS: 229-612-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR