7-Aminoenanthic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 7-Aminoheptanoic acid, 7-Aminoheptylic acid

H27519-03 H27519-06
AAH27519-03EA 128 USD
AAH27519-03 AAH27519-06 AAH27519-MD
7-Aminoenanthic acid 98%
7-Aminoenanthic acid

Purity: 98% CAS No.: 929-17-9 Molecular Formula: H2N(CH2)6CO2H Formula Weight: 145.19 Melting Point: 194-196° Beilstein Reference: 906887

Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
Melting Pt: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00008242
CAS Number: 929-17-9
EINECS: 213-197-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR