5-Methoxy-m-cresol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxy-5-methylanisol, 1-Hydroxy-3-methoxy-5-methylbenzene, 3-Methoxy-5-methylphenol, 3-Hydroxy-5-methoxytoluene

H27516-03 H27516-06
AAH27516-03EA 45.5 USD
AAH27516-03 AAH27516-06
5-Methoxy-m-cresol 97%
5-Methoxy-m-cresol

Purity: 97% CAS No.: 3209-13-0 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 60-66° MDL No.: MFCD00059261

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Melting Pt: 60…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00059261
CAS Number: 3209-13-0
EINECS: 221-716-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR