D-(-)-Isoleucine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3R)-(-)-Isoleucine, H-D-Ile-OH
H27488-03
H27488-MD
AAH27488-03EA
726
USD
AAH27488-03
AAH27488-MD
D-(-)-Isoleucine 98%
D-(-)-Isoleucine
Purity: 98%
CAS No.: 319-78-8
Molecular Formula: C6H13NO2
Formula Weight: 131.18
Melting Point: 265° subl.
Optical Rotation: -40° (c=5 in 5N HCl)
Formula:
H₃CCH₂CH(CH₃)CH(NH₂)COOH MW: 131.17 Boiling Pt: 235 Melting Pt: 98 (subl.) Density: 1.224 Storage Temperature: Ambient |
MDL Number:
MFCD00064221 CAS Number: 319-78-8 EINECS: 206-269-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...