2-Amino-3-nitrophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydroxy-6-nitroaniline
H27472-06
H27472-14
AAH27472-06EA
71.4
USD
AAH27472-06
AAH27472-14
2-Amino-3-nitrophenol 98%
2-Amino-3-nitrophenol
Purity: 98%
CAS No.: 603-85-0
Molecular Formula: C6H6N2O3
Formula Weight: 154.13
Melting Point: ca 212°
Beilstein Reference: 2804307
Formula:
C₆H₆N₂O₃ MW: 154.13 g/mol Melting Pt: ∼ 212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010875 CAS Number: 603-85-0 EINECS: 210-060-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...