2-Cyanophenylboronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Cyanobenzeneboronic acid pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 2-Cyanophenylboronic acid, pinacol ester

H27469-06 H27469-14
AAH27469-06EA 119 USD
AAH27469-06 AAH27469-14 AAH27469-03
2-Cyanophenylboronic acid pinacol ester 98%
2-Cyanophenylboronic acid pinacol ester

Purity: 98% CAS No.: 214360-48-2 Molecular Formula: C13H16BNO2 Formula Weight: 229.08 Melting Point: 88-90° MDL No.: MFCD04038747

Formula: C₁₃H₁₆BNO₂
MW: 229.09 g/mol
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD04038747
CAS Number: 214360-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR