1-Ethyl-3-methylimidazolium diethylphosphate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-ethyl-3-methylimidazolium diethylphosphate
H27400-06
H27400-14
AAH27400-06EA
52.3
USD
AAH27400-06
AAH27400-14
1-Ethyl-3-methylimidazolium diethylphosphate 98%
1-Ethyl-3-methylimidazolium diethylphosphate
Purity: 98%
CAS No.: 848641-69-0
Molecular Formula: C10H21N2O4P
Formula Weight: 264.26
Physical Form: Liquid
Density: 1.157
Appearance: Yellow to orange
Melting Point: 19-21°
UN No.: 3265
MDL No.: MFCD09953486
Formula:
C₁₀H₂₁N₂O₄P MW: 264.26 g/mol Melting Pt: 19…21 °C Density: 1.157 Storage Temperature: Ambient |
MDL Number:
MFCD09953486 CAS Number: 848641-69-0 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...