Potassium(4-methoxyphenyl)trifluoroborate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Methoxyphenyl)trifluoroboric acid potassium salt, Potassium (4-methoxyphenyl)trifluoroborate, Potassium trifluoro(4-methoxyphenyl)borate
H27384-03
H27384-06
AAH27384-03EA
56.6
USD
AAH27384-03
AAH27384-06
Potassium(4-methoxyphenyl)trifluoroborate 95%
Potassium(4-methoxyphenyl)trifluoroborate
Purity: 95%
CAS No.: 192863-36-8
Molecular Formula: C7H7BF3KO
Formula Weight: 214.04
Melting Point:
Formula:
C₇H₇BF₃KO MW: 214.04 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD04112717 CAS Number: 192863-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...