4-Aminopyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

H27367-06 H27367-MD H27367-03
AAH27367-06EA 513 USD
AAH27367-06 AAH27367-MD AAH27367-03
4-Aminopyrimidine 98%
4-Aminopyrimidine

Purity: 98% CAS No.: 591-54-8 Molecular Formula: C4H5N3 Formula Weight: 95.11 Melting Point: 147-152° MDL No.: MFCD00006112

Formula: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 147…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00006112
CAS Number: 591-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR