1-Pentyl-4-iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Iodopentylbenzene, 4-Pentyliodbenzol, 1-(4-Iodophenyl)pentane, 1-Iodo-4-pentylbenzene, 4-Pentyliodobenzene, 1-Iodo-4-n-pentylbenzene

H27356-03 H27356-09
AAH27356-03EA 35.4 USD
AAH27356-03 AAH27356-09
1-Pentyl-4-iodobenzene 98%
1-Pentyl-4-iodobenzene

Purity: 98% CAS No.: 85017-60-3 Molecular Formula: C11H15I Formula Weight: 274.14 Flash Point: 110°(230°F) Boiling Point: 88°/1mm MDL No.: MFCD00079767

Formula: C₁₁H₁₅I
MW: 274.14 g/mol
Boiling Pt: 88 °C/1 mm
Density: 1.38
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00079767
CAS Number: 85017-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR