2'-Aminoacetanilide 98%

Supplier: Thermo Scientific Chemicals
Warning

H27343-14 H27343-06
AAH27343-14EA 236 USD
AAH27343-14 AAH27343-06
2'-Aminoacetanilide 98%
2'-Aminoacetanilide

Purity: 98% CAS No.: 34801-09-7 Molecular Formula: C8H10N2O Formula Weight: 150.18 Melting Point: 133-137° MDL No.: MFCD00210388

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00210388
CAS Number: 34801-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR