1-Butyl-3-methylimidazolium methanesulfonate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H27336-18
H27336-06
AAH27336-18EA
172
USD
AAH27336-18
AAH27336-06
1-Butyl-3-methylimidazolium methanesulfonate 99%
1-Butyl-3-methylimidazolium methanesulphonate
Purity: 99%
CAS No.: 342789-81-5
Molecular Formula: C9H18N2O3S
Formula Weight: 234.32
Flash Point: 119°(246°F)
Melting Point: 75-80°
UN No.: 3261
MDL No.: MFCD06798173
Formula:
C₉H₁₈N₂O₃S Melting Pt: 75…80 °C Flash Pt: 119 °C (246 °F) Storage Temperature: Ambient |
MDL Number:
MFCD06798173 CAS Number: 342789-81-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...