2-Bromo-6-nitrophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27326-14
H27326-06
AAH27326-14EA
325
USD
AAH27326-14
AAH27326-06
2-Bromo-6-nitrophenol 97%
2-Bromo-6-nitrophenol
Purity: 97%
CAS No.: 13073-25-1
Molecular Formula: C6H4BrNO3
Formula Weight: 218.00
Melting Point: 66-69°
MDL No.: MFCD02683216
Formula:
C₆H₄BrNO₃ MW: 218.01 g/mol Melting Pt: 66…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683216 CAS Number: 13073-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...