4'-Chloro-2'-hydroxyacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27309-18
H27309-09
AAH27309-18EA
324
USD
AAH27309-18
AAH27309-09
4'-Chloro-2'-hydroxyacetophenone 97%
4'-Chloro-2'-hydroxyacetophenone
Purity: 97%
CAS No.: 6921-66-0
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Melting Point: 27-30°
MDL No.: MFCD00238557
Formula:
C₈H₇ClO₂ Melting Pt: 27…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00238557 CAS Number: 6921-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...