2-Bromoquinoline 96%

Supplier: Thermo Scientific Chemicals
Warning

H27302-06 H27302-14 H27302-03
AAH27302-06EA 194 USD
AAH27302-06 AAH27302-14 AAH27302-03
2-Bromoquinoline 96%
2-Bromoquinoline

Purity: 96% CAS No.: 2005-43-8 Molecular Formula: C9H6BrN Formula Weight: 208.06 MDL No.: MFCD00234480

Formula: C₉H₆BrN
Storage Temperature: Ambient
MDL Number: MFCD00234480
CAS Number: 2005-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR