4-Bromo-2-chloropyridine 94%

Supplier: Thermo Scientific Chemicals
Warning

H27299-06 H27299-14
AAH27299-06EA 34.6 USD
AAH27299-06 AAH27299-14
4-Bromo-2-chloropyridine 94%
4-Bromo-2-chloropyridine

Purity: 94% CAS No.: 73583-37-6 Molecular Formula: C5H3BrClN Formula Weight: 192.44 Flash Point: 104°(219°F) Melting Point: 18-20° Boiling Point: 70°/3mm MDL No.: MFCD03840756

Formula: C₅H₃BrClN
MW: 192.44 g/mol
Boiling Pt: 70 °C/3 mm
Melting Pt: 18…20 °C
Density: 1.734
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD03840756
CAS Number: 73583-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR