Methyl-3-bromo-4-methoxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-bromo-4-methoxybenzoate, 3-Bromo-4-methoxybenzoic acid methyl ester

H27297-09 H27297-03
AAH27297-09EA 139 USD
AAH27297-09 AAH27297-03
Methyl-3-bromo-4-methoxybenzoate 98%
Methyl-3-bromo-4-methoxybenzoate

Purity: 98% CAS No.: 35450-37-4 Molecular Formula: C9H9BrO3 Formula Weight: 245.08 Melting Point: 95-98° MDL No.: MFCD00210463

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00210463
CAS Number: 35450-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR