2-Amino-5-bromopyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromopyrimidin-2-ylamine, 5-Bromopyrimidin-2-amine

H27291-03 H27291-06
AAH27291-03EA 22.7 USD
AAH27291-03 AAH27291-06
2-Amino-5-bromopyrimidine 97%
2-Amino-5-bromopyrimidine

Purity: 97% CAS No.: 7752-82-1 Molecular Formula: C4H4BrN3 Formula Weight: 174.00 Melting Point: 232-234° MDL No.: MFCD00012341

Formula: C₄H₄BrN₃
MW: 174 g/mol
Melting Pt: 232…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00012341
CAS Number: 7752-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR