1-Hexyl-3-methylimidazolium hexafluorophosphate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Hexyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)
H27272-06
H27272-18
AAH27272-06EA
103
USD
AAH27272-06
AAH27272-18
1-Hexyl-3-methylimidazolium hexafluorophosphate 99%
1-Hexyl-3-methylimidazolium hexafluorophosphate
Purity: 99%
CAS No.: 304680-35-1
Molecular Formula: C10H19F6N2P
Formula Weight: 312.24
Refractive Index: 1.419
Formula:
C₁₀H₁₉F₆N₂P MW: 312.24 g/mol Density: 1.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03093296 CAS Number: 304680-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...