3-Bromo-4-nitrobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H27268-06 H27268-03
AAH27268-06EA 174 USD
AAH27268-06 AAH27268-03
3-Bromo-4-nitrobenzoic acid 97%
3-Bromo-4-nitrobenzoic acid

Purity: 97% CAS No.: 101420-81-9 Molecular Formula: C7H4BrNO4 Formula Weight: 246.02 Melting Point: 200-204° MDL No.: MFCD04117949

Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Melting Pt: 200…204 °C
Storage Temperature: Ambient
MDL Number: MFCD04117949
CAS Number: 101420-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR