4-Bromo-3-chlorobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27252-06
H27252-14
AAH27252-06EA
70.7
USD
AAH27252-06
AAH27252-14
4-Bromo-3-chlorobenzoic acid 97%
4-Bromo-3-chlorobenzoic acid
Purity: 97%
CAS No.: 25118-59-6
Molecular Formula: C7H4BrClO2
Formula Weight: 235.47
Odor: Odorless
Melting Point: 220-225°
MDL No.: MFCD08276864
Formula:
C₇H₄BrClO₂ MW: 235.46 g/mol Melting Pt: 220…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276864 CAS Number: 25118-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...