1-Ethyl-3-methylimidazolium hydrogen sulfate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Ethyl-3-methylimidazolium Hydrogen sulphate

H27232-18 H27232-30
AAH27232-18EA 131 USD
AAH27232-18 AAH27232-30
1-Ethyl-3-methylimidazolium hydrogen sulfate 98%
1-Ethyl-3-methylimidazolium hydrogen sulphate

Purity: 98% CAS No.: 412009-61-1 Molecular Formula: C6H12N2O4S Formula Weight: 208.24 UN No.: 3265 MDL No.: MFCD06798195

Formula: C₆H₁₂N₂O₄S
MW: 208.24 g/mol
Density: 1.367
Storage Temperature: Ambient
MDL Number: MFCD06798195
CAS Number: 412009-61-1
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR