(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(4-Hydroxy-3,5-dimethoxyphenyl)acetic acid, 4-Hydroxy-3,5-dimethoxyphenylacetic acid
H27231-06
H27231-03
AAH27231-06EA
114
USD
AAH27231-06
AAH27231-03
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid 97%
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid
Purity: 97%
CAS No.: 4385-56-2
Molecular Formula: C10H12O5
Formula Weight: 212.21
Melting Point: 127-131°
Beilstein Reference: 2851658
Formula:
C₁₀H₁₂O₅ Melting Pt: 127…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016834 CAS Number: 4385-56-2 EINECS: 224-494-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...