2-Bromo-5-hydroxypyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromo-5-pyridinol, 6-Bromopyridin-3-ol
H27228-03
H27228-06
AAH27228-03EA
98.8
USD
AAH27228-03
AAH27228-06
2-Bromo-5-hydroxypyridine 95%
2-Bromo-5-hydroxypyridine
Purity: 95%
CAS No.: 55717-45-8
Molecular Formula: C5H4BrNO
Formula Weight: 174.00
Melting Point: 133-136°
MDL No.: MFCD04114184
Formula:
C₅H₄BrNO Melting Pt: 133…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114184 CAS Number: 55717-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...