1,2,3,4,5-Pentaphenylcyclopentadiene 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene, Cyclopenta-1,3-diene-1,2,3,4,5-pentaylpentabenzene, Pentaphenylcyclopentadiene

H27223-03 H27223-06
AAH27223-03EA 122 USD
AAH27223-03 AAH27223-06 AAH27223-MD
1,2,3,4,5-Pentaphenylcyclopentadiene 99%
1,2,3,4,5-Pentaphenylcyclopentadiene

Purity: 99% CAS No.: 2519-10-0 Molecular Formula: C35H26 Formula Weight: 446.59 Melting Point: 254-257° MDL No.: MFCD09909572

Formula: C₃₅H₂₆
MW: 446.59 g/mol
Melting Pt: 254…257 °C
Storage Temperature: Ambient
MDL Number: MFCD00012263
CAS Number: 2519-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR