2-Bromo-5-thiazolecarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27203-06
H27203-03
AAH27203-06EA
224
USD
AAH27203-06
AAH27203-03
2-Bromo-5-thiazolecarboxylic acid 97%
2-Bromo-5-thiazolecarboxylic acid
Purity: 97%
CAS No.: 54045-76-0
Molecular Formula: C4H2BrNO2S
Formula Weight: 208.03
Melting Point: 181-183°
MDL No.: MFCD04115730
Formula:
C₄H₂BrNO₂S Melting Pt: 181…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115730 CAS Number: 54045-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...