1-Butyl-3-methylimidazolium bromide 99%

Supplier: Thermo Scientific Chemicals
Warning

H27201-06 H27201-18
AAH27201-06EA 31.3 USD
AAH27201-06 AAAH27201-18
1-Butyl-3-methylimidazolium bromide 99%
1-Butyl-3-methylimidazolium bromide

Purity: 99% CAS No.: 85100-77-2 Molecular Formula: C8H15BrN2 Formula Weight: 219.12 Melting Point: 65-75° MDL No.: MFCD03427611

Formula: C₈H₁₅BrN₂
Melting Pt: 65…75 °C
Storage Temperature: Ambient
MDL Number: MFCD03427611
CAS Number: 85100-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR